Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

748358

Sigma-Aldrich

2-(Di-tert-butylphosphino)ethylamine

10 wt. % in THF

Synonym(s):

2-(Di-tert-butylphosphino)ethylamine solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H24NP
CAS Number:
Molecular Weight:
189.28
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

concentration

10 wt. % in THF

refractive index

n20/D 1.420

density

0.879 g/mL at 25 °C

functional group

phosphine

SMILES string

CC(C)(C)P(CCN)C(C)(C)C

InChI

1S/C10H24NP/c1-9(2,3)12(8-7-11)10(4,5)6/h7-8,11H2,1-6H3

InChI key

DZDGRVHIUYIMDS-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk_germany

WGK 3

flash_point_f

-5.1 °F

flash_point_c

-20.6 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service