Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

748293

Sigma-Aldrich

(R)-1-(Diphenylphosphino)-3,3-dimethyl-2-butylamine

97%

Synonym(s):

(R)-1-(Diphenylphosphino)-3,3-dimethylbutan-2-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24NP
Molecular Weight:
285.36
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

assay

97%

form

liquid

reaction suitability

reagent type: ligand

refractive index

n20/D 1.589

density

1.026 g/mL at 25 °C

functional group

phosphine

SMILES string

CC(C)(C)[C@@H](N)CP(c1ccccc1)c2ccccc2

InChI

1S/C18H24NP/c1-18(2,3)17(19)14-20(15-10-6-4-7-11-15)16-12-8-5-9-13-16/h4-13,17H,14,19H2,1-3H3/t17-/m0/s1

Inchi Key

SGBNMEAXYLJQRV-KRWDZBQOSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

230.0 °F

flash_point_c

110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service