Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

747211

Sigma-Aldrich

Merocyanine dye, HB194

Synonym(s):

2-((Z)-2-((E)-2-(1,1-Dimethyl-5,6-dihydro-4H-pyrrolo[3,2,1-ij]quinolin-2(1H)-ylidene)ethylidene)-3-oxo-2,3-dihydro-1H-inden-1-ylidene)malononitrile, 2-[(Z)-2-[(E)-2-(5,6-Dihydro-1,1-dimethyl-4H-pyrrolo[3,2,1-ij]quinolin-2(1H)-ylidene)ethylidene]-2,3-dihydro-3-oxo-1H-inden-1-ylidene]propanedinitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H21N3O
Molecular Weight:
403.48
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

mp

288-290 °C

λmax

~578 nm in dichloromethane

SMILES string

O=C(C1=C2C=CC=C1)/C(C2=C(C#N)C#N)=C\C=C3C(C)(C)C4=C5C(CCCN5/3)=CC=C4

InChI

1S/C27H21N3O/c1-27(2)22-11-5-7-17-8-6-14-30(25(17)22)23(27)13-12-21-24(18(15-28)16-29)19-9-3-4-10-20(19)26(21)31/h3-5,7,9-13H,6,8,14H2,1-2H3/b21-12-,23-13+

InChI key

XCFYTQCAMLKFSO-ABLMNQNHSA-N

General description

Merocyanine dye, HB194 belongs to the class of absorption materials that are majorly used as donor molecules in organic electronic devices. It has a high absorption coefficient and good polarizing properties, which allow it to be utilized as a photorefractive material.[1][2]

Application

VAC-processed single high-efficiency Small-BHJ solar cell based on the MC dye
OPV Device Structure: ITO/MoO3/HB194: C60/BPhen/Al
  • JSC = 13.0 mA/cm2
  • VOC = 0.96 V
  • FF = 0.49
  • PCE = 6.1%

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Steinmann; Vera;
Advanced Energy Materials, 1(5), 888-893 (2011)
Direct comparison of highly efficient solution-and vacuum-processed organic solar cells based on merocyanine dyes.
Kronenberg NM, et al.
Advanced Materials, 22(37), 4193-4197 (2010)
Tailored merocyanine dyes for solution-processed BHJ solar cells.
Burckstummer H, et al.
Journal of Materials Chemistry, 20(2), 240-243 (2010)

Articles

While dye sensitization as the basis for color photography has been accepted for a very long time,1 attempts to use this principle for the conversion of solar light to electricity generally had resulted only in very low photocurrents, below 100 nA/cm

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service