Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

746916

Sigma-Aldrich

Polystyrene, azide terminated

average Mn 15,000, PDI ≤1.3

Synonym(s):

PS N3 terminated, clickable PS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C7H12N3O2[CH2CH(C6H5)]nH
UNSPSC Code:
12352100

form

powder

Quality Level

mol wt

average Mn 15,000

PDI

≤1.3

Looking for similar products? Visit Product Comparison Guide

Application

This is a well-defined polystyrene with an azide end group functionality. Azido groups are commonly used in Cu-mediated ligation (azide-alkyne click chemistry).

Legal Information

Sold for research purposes only and subject to field restriction: see www.sigmaaldrich.com/raftlicense. Patents: 7; 714; 075; 7; 250; 479; 7; 666; 962; 6; 642; 318; 6; 747; 111; W2010/8356.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service