Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

746665

Sigma-Aldrich

Gold nanoparticles

250 nm diameter, stabilized suspension in 0.1 mM PBS, reactant free

Synonym(s):

Gold nanodispersion, Au NP, Gold colloid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

nanoparticles
suspension

concentration

~7.1E+8 particles/mL

OD

1

diameter

250 nm

storage temp.

2-8°C

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Product of CytoDiagnostics, Inc.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Graphene is a unique two-dimensional (2D) structure of monolayer carbon atoms packed into a dense honeycomb crystal that has attracted great interest due to its diverse and fascinating properties.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service