Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

746606

Sigma-Aldrich

D358 Dye

95%

Synonym(s):

5-[3-(Carboxymethyl)-5-[[4-[4-(2,2-diphenylethenyl)phenyl]-1,2,3,3a,4,8b-hexahydrocyclopent[b]indol-7-yl]methylene]-4-oxo-2-thiazoli dinylidene]-4-oxo-2-thioxo-3-thiazolidinedodecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H53N3O6S3
CAS Number:
Molecular Weight:
912.19
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

95%

form

powder

λmax

532 nm in DMF

SMILES string

S=C(S/C1=C2N(CC(O)=O)C(/C(S\2)=C\C(C=C3)=CC4=C3N(C5=CC=C(C=C(C6=CC=CC=C6)C7=CC=CC=C7)C=C5)C8C4CCC8)=O)N(CCCCCCCCCCCC(O)=O)C1=O

InChI

1S/C52H53N3O6S3/c56-46(57)23-14-6-4-2-1-3-5-7-15-30-53-50(61)48(64-52(53)62)51-54(34-47(58)59)49(60)45(63-51)33-36-26-29-44-42(32-36)40-21-16-22-43(40)55(44)39-27-24-35(25-28-39)31-41(37-17-10-8-11-18-37)38-19-12-9-13-20-38/h8-13,17-20,24-29,31-33,40,43H,1-7,14-16,21-23,30,34H2,(H,56,57)(H,58,59)/b45-33+,51-48+

InChI key

HXSDFCKKOUCOIZ-DOESQJKTSA-N

Application

Photosensitive Red-Violet dye having excellent photoelectric conversion efficiency as a metal-free dye.
Efficiency of DSSC device - η > 6.7%
Cell area: 0.25 cm2 (0.5 cm × 0.5 cm)
TiO2 Thickness: 5 μm
TiO2: screen printing
Electrolyte: 0.1 M LiI; 0.05 M I2; 0.6 M Dimethyl propylimidazolium
Iodide; 0.05 M t-Butylpyridine in 3-Methoxypropionitrile
Light source: AM1.5
Counter Electrode: Pt/Cr spatter glass
Dye adsorption: 30 °C/ in 3 hr. (t-BuOH/acetonitrile soln.)

Legal Information

Product of Mitsubishi Paper Mills; Ltd.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Premaratne; K.;
J. Photochem. Photobiol., A, 229(1), 29-32 (2012)

Articles

Dye-sensitized solar cells (DSCs) are 3rd generation solar cells combining the promise of high efficiency with low production costs.

While dye sensitization as the basis for color photography has been accepted for a very long time,1 attempts to use this principle for the conversion of solar light to electricity generally had resulted only in very low photocurrents, below 100 nA/cm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service