746312
Zinc monoantimonide
-80 mesh, 99.9% trace metals basis
Synonym(s):
Zinc antimonide
About This Item
assay
99.9% trace metals basis
form
powder
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
particle size
-80 mesh
greener alternative category
, Enabling
SMILES string
[Zn+3].[Sb-3]
InChI
1S/Sb.Zn/q-3;+3
InChI key
ADMLQACAMWIQCT-UHFFFAOYSA-N
General description
Application
In recent years ZnSb is also used as anode material for lithium ion batteries.
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service