Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

746096

Sigma-Aldrich

4-(Hydroxymethyl)-2-methylthiazole

Synonym(s):

(2-Methyl-1,3-thiazol-4-yl)methanol, (2-Methylthiazol-4-yl)methanol, 2-Methyl-4-thiazolemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7NOS
CAS Number:
Molecular Weight:
129.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

form

solid

Quality Level

mp

32-37 °C

SMILES string

OCC1=CSC(C)=N1

InChI

1S/C5H7NOS/c1-4-6-5(2-7)3-8-4/h3,7H,2H2,1H3

InChI key

KRCIFTBSQKDYMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service