Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

745995

Sigma-Aldrich

Zinc antimonide

greener alternative

powder, -80 mesh, 99.9% trace metals basis

Synonym(s):

Zinc tetraantimonide, Zn4Sb3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn4Sb3
CAS Number:
Molecular Weight:
626.84
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

assay

99.9% trace metals basis

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

-80 mesh

greener alternative category

SMILES string

[Zn+2].[Zn+2].[Zn+2].[Zn+].[Sb-3].[Sb-3].[Sb-3]

InChI

1S/3Sb.4Zn/q3*-3;+1;3*+2

InChI key

SZFDATULCDXOQB-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.
Zinc antimonide (Zn4Sb3) is a p-type high-performance thermoelectric (TE) material, which is of interest because of its low cost, relatively large abundance, low toxicity, and low thermal conductivity. It can be used as an anode for lithium-ion batteries.[1][2][3][4]

Application

Zn4Sb3 is one of the most efficient thermoelectric material mainly due to its extraordinarily low thermal conductivity in conjunction with the electronic structure of a heavily doped semiconductor. It can be doped with different elements like Pb, Sb etc to further enhance its thermopower.

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johanna Nylén et al.
Journal of the American Chemical Society, 126(50), 16306-16307 (2004-12-17)
beta-Zn4Sb3 is an outstanding thermoelectric material mainly due to its extraordinarily low thermal conductivity, which is similar to that of glasses. Recently it was proposed that interstitial Zn atoms are responsible for this peculiar behavior. Here we report on the
Zinc antimonide thin films prepared by ion beam sputtering deposition using ternary layers annealing method
Zheng Z, et al.
Journal of alloys and compounds, 594(13), 122-126 (2014)
Electrochemical properties of Zn4Sb3 as anode materials for lithium-ion batteries
Zhao X, et al.
Journal of Materials Science Letters, 19(10), 851-853 (2000)
Tang,
Journal of Electronic Materials, 41(6), 1091-1091 (2012)
G Jeffrey Snyder et al.
Nature materials, 3(7), 458-463 (2004-06-29)
By converting waste heat into electricity, thermoelectric generators could be an important part of the solution to today's energy challenges. The compound Zn(4)Sb(3) is one of the most efficient thermoelectric materials known. Its high efficiency results from an extraordinarily low

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service