Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

745987

Sigma-Aldrich

Cobalt triantimonide

greener alternative

powder, -80 mesh, 99.9% trace metals basis

Synonym(s):

CoSb3, Cobalt antimonide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CoSb3
CAS Number:
Molecular Weight:
424.21
UNSPSC Code:
26111700
NACRES:
NA.23

assay

99.9% trace metals basis

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

-80 mesh

mp

>400 °C

greener alternative category

InChI

1S/Co.3Sb

InChI key

RSEZDFOSLUSNIU-UHFFFAOYSA-N

General description

Cobalt triantimonide (CoSb3) is a thermoelectric (TE) material with a power factor, PF of 30 μW cm−1 and a Seebeck coefficient, S of 200 μV K−1. It is a covalent semiconductor material, which has a body-centered cubic (BCC) structure.[1][2]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Cobalt antimonide is a skutterudite which has been widely studied for its thermoelectric properties. It can be doped with different elements to further increase its thermopower. By varying the dopant element it can be tuned to exhibit either p- or n-type semiconductor properties.

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and Characterization of Electrochemical Deposition Cobalt Triantimonide (CoSb 3) Thick Film: Effects of Polyvinyl Alcohol (PVA) as an Additive
Sabran NS, et al.
Journal of Electronic Materials, 48(8), 5003-5011 (2019)
Godart,
J. Alloy Compounds, 55-55 (2013)
Electronic Structure and Phase Stability of Yb-Filled CoSb3 Skutterudite Thermoelectrics from First-Principles
Isaacs EB and Wolverton C
Chemistry of Materials, 31(16), 6154-6162 (2019)
Caillat, T.;
Journal of Applied Physics, 80(8), 4442-4442 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service