Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

745901

Sigma-Aldrich

PBTTPD

Synonym(s):

Poly[[5-(2-ethylhexyl)-5,6-dihydro-4,6-dioxo-4H-thieno[3,4-c]pyrrole-1,3-diyl](4,4′-didodecyl[2,2′-bithiophene]-5,5′-diyl)]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C46H69NO2S3)n
CAS Number:
UNSPSC Code:
12352103
NACRES:
NA.23

description

Band gap: 1.82 eV

form

solid

mol wt

average Mn 3,500-20,000 g/mol

λmax

572 nm (thin film)

orbital energy

HOMO -5.56 eV 
LUMO -3.1 eV 

PDI

1.2‑2.5

General description

HOMO: -5.56 eV
LUMO: -3.10 eV
UV absorption: λmax = 572 nm;Eg = 1.82 eV
Solubility: This polymer can be dissolved easily in hot chloroform;hot chlorobenzene;hot dichlorobenzene
PBTTPD is a donor-acceptor based high crystalline conducting polymer. It has thieno[3,4-c]pyrrole-4,6-dione (TPD), which acts as the electron-withdrawing group. Bi(dodecyl)thiophene based donor groups facilitate the polymeric unit with crystallinity and low HOMO energy levels. It shows good hole mobility and open circuit voltage (VOC).[1][2]

Application

  • High-Efficiency Organic Solar Cells (OPVs)
  • Power conversion efficiency (PCE): 7.3%
  • Open circuit voltage (Voc): 0.92 V
  • Short circuit current density (Jsc): 13.1 mA/cm2
  • Fill factor (FF): 0.61
PBTTPD is majorly used as a conjugating polymer in the fabrication of organic solar cells (OSCs) and organic photovoltaic cells (OPVs). It has a power conversion efficiency (PCE) of 7.3% and a VOC of 0.92V.[3][4]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural Evolution of Crystalline Conjugated Polymer/Fullerene Domains from Solution to the Solid State in the Presence and Absence of an Additive
Su Y, et al.
The Journal of Physical Chemistry C, 119(6), 3408-3417 (2015)
Crystalline donor-acceptor conjugated polymers for bulk heterojunction photovoltaics
Jiang J, et al.
Journal of Material Chemistry A, 1(14), 4415-4422 (2013)
A thieno [3, 4-c] pyrrole-4, 6-dione-based donor- acceptor polymer exhibiting high crystallinity for photovoltaic applications
Yuan M, et al.
Macromolecules, 43(17), 6936-6938 (2010)
Improving device efficiency of polymer/fullerene bulk heterojunction solar cells through enhanced crystallinity and reduced grain boundaries induced by solvent additives.
Ming-Shin Su et al.
Advanced materials (Deerfield Beach, Fla.), 23(29), 3315-3319 (2011-06-15)

Articles

The development of high-performance conjugated organic molecules and polymers has received widespread attention in industrial and academic research.

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service