Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

745804

Sigma-Aldrich

Bismuth antimonide

greener alternative

powder, -80 mesh, 99.9% trace metals basis

Synonym(s):

BiSb

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BiSb
CAS Number:
Molecular Weight:
330.74
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

assay

99.9% trace metals basis

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

-80 mesh

greener alternative category

SMILES string

[Sb]#[Bi]

InChI

1S/Bi.Sb

InChI key

PEEDYJQEMCKDDX-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Bismuth antimonide (BiSb) can be used in the formation of a thin film based thermopile for the fabrication of heat power sensors for biochemical applications. It can also be used as an active element in the designing of a photo-thermoelectric sensor.[1][2]
Skutterudites are a promising class of thermoelectric materials which can be doped with other elements like rare earth elements to decrease thermal conductivity without reducing the electrical conductivity, hence increasing the thermopower.

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new micro-fluid chip calorimeter for biochemical applications
Lerchner J, et al.
Thermochimica Acta, 445(2), 144-150 (2006)
Lee;
J. Alloy Compounds, 553, 343-343 (2013)
A YBaCuO-BiSb photo-thermoelectric sensor: mathematical modeling of normalised detectivity
Kaila MM
Physica C, 406(3-4), 205-209 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service