745545
2-Bromo-4-ethylpyridine
96%
Synonym(s):
4-Ethyl-2-bromopyridine
About This Item
Recommended Products
assay
96%
form
liquid
bp
239.5-20.0 °C (predicted)
SMILES string
CCc1ccnc(Br)c1
InChI
1S/C7H8BrN/c1-2-6-3-4-9-7(8)5-6/h3-5H,2H2,1H3
InChI key
JMDIWBIWPAAWHP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service