745324
Bis(5H-dibenzo[a,d]cyclohepten-5-yl)phenylphosphine
≥96.5% (elemental analysis)
Synonym(s):
Bis(5H-dibenzo[a,d]cyclohepten-5-yl)phenylphosphane, trop2-PPh
About This Item
assay
≥96.5% (elemental analysis)
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
loss
≤2.0% loss on drying
functional group
phosphine
SMILES string
c1ccc(cc1)P(C2c3ccccc3C=Cc4ccccc24)C5c6ccccc6C=Cc7ccccc57
InChI
1S/C36H27P/c1-2-16-30(17-3-1)37(35-31-18-8-4-12-26(31)22-23-27-13-5-9-19-32(27)35)36-33-20-10-6-14-28(33)24-25-29-15-7-11-21-34(29)36/h1-25,35-36H
InChI key
FPKRJQIFIQXPKV-UHFFFAOYSA-N
Packaging
hcodes
Hazard Classifications
Aquatic Chronic 4
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service