Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

745138

Sigma-Aldrich

(1S,2R)-cis-N-Boc-1,2-cyclopentanediamine

≥97% (TLC)

Synonym(s):

(1R,2S)-2-(Boc-amino)cyclopentylamine, (1S)-cis-N-Boc-1,2-diaminocyclopentane, tert-Butyl ((1S,2R)-2-aminocyclopentyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20N2O2
CAS Number:
Molecular Weight:
200.28
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

assay

≥97% (TLC)

form

solid

optical purity

enantiomeric excess: ≥95.0%

SMILES string

N[C@H]1[C@@H](NC(OC(C)(C)C)=O)CCC1

InChI

1S/C10H20N2O2/c1-10(2,3)14-9(13)12-8-6-4-5-7(8)11/h7-8H,4-6,11H2,1-3H3,(H,12,13)/t7-,8+/m1/s1

InChI key

PAXDIBGWURAVIH-SFYZADRCSA-N

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z Trobonjaca et al.
Journal of immunology (Baltimore, Md. : 1950), 167(3), 1413-1422 (2001-07-24)
A prominent subset of the hepatic innate immune system is alpha-galactosylceramide (alphaGalCer)-reactive, (CD4(+) and CD4(-)CD8(-)) CD1d-restricted NKT cells. We investigated in C57BL/6 (B6) mice which hepatic cell type stimulates hepatic NKT cell activation. Surface expression of CD1d but not CD40
U Sandhu et al.
Nucleic acids research, 39(1), e1-e1 (2010-10-12)
Recombinant mouse strains that harbor tightly controlled transgene expression proved to be indispensible tools to elucidate gene function. Different strategies have been employed to achieve controlled induction of the transgene. However, many models are accompanied by a considerable level of
Zlatko Trobonjaca et al.
Journal of immunology (Baltimore, Md. : 1950), 168(8), 3763-3770 (2002-04-09)
Dendritic cell (DC)-dependent activation of liver NKT cells triggered by a single i.v. injection of a low dose (10-100 ng/mouse) of alpha-galactosyl ceramide (alphaGalCer) into mice induces liver injury. This response is particularly evident in HBs-tg B6 mice that express
Reinhold Schirmbeck et al.
European journal of immunology, 33(12), 3342-3352 (2003-11-25)
Processing exogenous hepatitis B surface antigen (HBsAg) of the hepatitis B virus (HBV) generates the K(b)-binding S(208-215) epitope 1; processing endogenous HBsAg generates the K(b)-binding S(190-197) epitope 2. Cross-reactive CD8(+) T cell responses were primed to epitope 1 but not
Petra Riedl et al.
Journal of immunology (Baltimore, Md. : 1950), 183(1), 370-380 (2009-06-23)
Immunodominance limits the TCR diversity of specific antiviral CD8 T cell responses elicited by vaccination or infection. To prime multispecific T cell responses, we constructed DNA vaccines that coexpress chimeric, multidomain Ags (with CD8 T cell-defined epitopes of the hepatitis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service