Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

745103

Sigma-Aldrich

(2R)-N-[(1R)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-2-pyrrolidinecarboxamide

≥98.0% (HPLC)

Synonym(s):

(R)-N-((R)-1-Hydroxy-4-methyl-1,1-diphenylpentan-2-yl)pyrrolidine-2-carboxamide, N-[(1R)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-D-prolinamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H30N2O2
CAS Number:
Molecular Weight:
366.50
Beilstein/REAXYS Number:
19106160
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

≥98.0% (HPLC)

form

powder

optical activity

[α]/D 50.9±3°, c = 1 in chloroform

mp

194 °C±3 °C

functional group

amide
hydroxyl
phenyl

SMILES string

CC(C)C[C@@H](NC(=O)[C@H]1CCCN1)C(O)(c2ccccc2)c3ccccc3

InChI

1S/C23H30N2O2/c1-17(2)16-21(25-22(26)20-14-9-15-24-20)23(27,18-10-5-3-6-11-18)19-12-7-4-8-13-19/h3-8,10-13,17,20-21,24,27H,9,14-16H2,1-2H3,(H,25,26)/t20-,21-/m1/s1

InChI key

PWBULUUPVDXEQS-NHCUHLMSSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

C-NMR and H-NMR conforms to structure

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monika Raj et al.
Organic letters, 8(18), 4097-4099 (2006-08-25)
We have demonstrated that a new class of l-proline-based organic compounds catalyzed the direct aldol reaction between aldehydes and acetone to provide beta-hydroxy ketones in good yields. The reaction is efficient, and 5-10 mol % of the catalyst and excellent
Direction of Kinetically versus Thermodynamically Controlled Organocatalysis and Its Application in Chemoenzymatic Synthesis
Rulli, G., et al.
Angewandte Chemie (International Edition in English), 50, 1-5 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service