Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

744913

Sigma-Aldrich

Di(1-adamantyl)(1-naphthoyl)phosphine

≥98% (HPLC)

Synonym(s):

Ad-BellGiphos-1-Nap

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H37OP
CAS Number:
Molecular Weight:
456.60
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

≥98% (HPLC)

SMILES string

O=C(P(C12CC3CC(CC(C3)C1)C2)C45CC6CC(CC(C6)C4)C5)c7cccc8ccccc78

InChI

1S/C31H37OP/c32-29(28-7-3-5-26-4-1-2-6-27(26)28)33(30-14-20-8-21(15-30)10-22(9-20)16-30)31-17-23-11-24(18-31)13-25(12-23)19-31/h1-7,20-25H,8-19H2

InChI key

BJOGEGHLGNQBBZ-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service