Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

744875

Sigma-Aldrich

Glass microreactor HTM V-type X-WÜ

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NA.22
Pricing and availability is not currently available.

General description

  • Borosilicate glass.
  • Dimensions 89 x 53 mm
  • Basic design element ST, Reagents: A1, A2, product: P, Parallel Channel Tempering WÜ
  • Connection: glass internal screw 1/4" UNF 28
  • Heat exchanger (WÜ) connections: hollow screws
  • Internal volume: 0,06 ml,
  • Permitted particle size : 50 μm,
  • Flow rate of water by 2(6) bar: 30 ml/min (70 ml/min).
  • Max pressure: 10 bar

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service