Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

744654

Sigma-Aldrich

2-Propynyl 2-acetamido-2-deoxy-α-D-glucopyranoside solution

~10 % (w/w) in methanol, ≥85% (HPLC)

Synonym(s):

2-Propynyl 2-acetamido-2-deoxy-α-D-glucoside solution, Propargyl 2-acetamido-2-deoxy-α-D-glucoside solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17NO6
CAS Number:
Molecular Weight:
259.26
Beilstein/REAXYS Number:
8996469
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

assay

≥85% (HPLC)

form

solution

concentration

~10 % (w/w) in methanol

storage temp.

2-8°C

SMILES string

CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@@H]1OCC#C

InChI

1S/C11H17NO6/c1-3-4-17-11-8(12-6(2)14)10(16)9(15)7(5-13)18-11/h1,7-11,13,15-16H,4-5H2,2H3,(H,12,14)/t7-,8-,9-,10-,11+/m1/s1

InChI key

LNFVQZQJEGUHLW-ILAIQSSSSA-N

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service