Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

744557

Sigma-Aldrich

5H-Dibenzo[a,d]cyclohepten-5-ylamine

95%

Synonym(s):

5-Amino-5H-dibenzo[a,d]cycloheptene, 5H-Dibenzo[a,d]cyclohepten-5-amine, trop-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13N
CAS Number:
Molecular Weight:
207.27
Beilstein/REAXYS Number:
2722512
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

≥95% (HPLC)
95%

form

powder

storage temp.

2-8°C

SMILES string

NC1c2ccccc2C=Cc3ccccc13

InChI

1S/C15H13N/c16-15-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)15/h1-10,15H,16H2

InChI key

HKLYXHWNPAGMMW-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Skull and crossbonesEnvironment

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalyzed Dehydrogenative Coupling of Primary Alcohols with Water, Methanol, or Amines
Zweifel, T., et al
Angewandte Chemie (International Edition in English), 49, 2009-2009 (2009)
Electromeric rhodium radical complexes.
Florian Frank Puschmann et al.
Angewandte Chemie (International ed. in English), 49(2), 385-389 (2009-12-04)
Torsten Büttner et al.
Chemical communications (Cambridge, England), 24(24), 2820-2821 (2004-12-16)
A combination of the rigid bis(5H-dibenzo[a,d]cyclohepten-5-yl)amine (trop(2)NH) and 5-amino-5H-dibenzo[a,d]cycloheptene (tropNH(2)) ligand allowed the synthesis of the stable pentacoordinated 18 electron amine olefin rhodium(i) complex [Rh(trop(2)NH)(tropNH(2))]O(3)SCF(3)(); this complex can be cleanly deprotonated [pK(a)(DMSO) = 20.6(1)] to the corresponding amide [Rh(trop(2)N)(tropNH(2))](6) which
Chiral Rhodium(I) and Iridium(I) Amino-Olefin Complexes:? pKa, N-H Bond Dissociation Energy, and Catalytic Transfer Hydrogenation
Maire, P., et al.
Organometallics, 24, 3207-3218 (2005)
Ethanol as hydrogen donor: highly efficient transfer hydrogenations with rhodium(I) amides.
Theo Zweifel et al.
Angewandte Chemie (International ed. in English), 47(17), 3245-3249 (2008-03-19)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service