Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

744530

Sigma-Aldrich

2-(2-Azidoethoxy)ethyl 2-acetamido-2-deoxy-α-D-glucopyranoside solution

~10 % (w/w) in methanol, ≥85% (HPLC)

Synonym(s):

2-(2-Azidoethoxy)ethyl 2-acetamido-2-deoxy-α-D-glucoside solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22N4O7
CAS Number:
Molecular Weight:
334.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥85% (HPLC)

form

solution

reaction suitability

reaction type: click chemistry

concentration

~10 % (w/w) in methanol

storage temp.

2-8°C

SMILES string

CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@@H]1OCCOCCN=[N+]=[N-]

InChI

1S/C12H22N4O7/c1-7(18)15-9-11(20)10(19)8(6-17)23-12(9)22-5-4-21-3-2-14-16-13/h8-12,17,19-20H,2-6H2,1H3,(H,15,18)/t8-,9-,10-,11-,12+/m1/s1

InChI key

YIYBKMXLQSWSPH-OOCWMUITSA-N

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

-27.4 °F

flash_point_c

-33 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service