Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

744522

Sigma-Aldrich

N-(Dichlorophosphino)dibenzo[b,f]azepine

95%

Synonym(s):

N-(Dichlorophosphanyl)dibenzo[b,f]azepine, P-5H-Dibenz[b,f]azepin-5-ylphosphonous dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10Cl2NP
CAS Number:
Molecular Weight:
294.12
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

assay

≥95.0% (CHN)
95%

form

powder

composition

carbon content, 56.9-57.7 wt. %
nitrogen content, 4.5-5.0 wt. %

reaction suitability

reagent type: ligand

functional group

phosphine

SMILES string

ClP(Cl)N1c2ccccc2C=Cc3ccccc13

InChI

1S/C14H10Cl2NP/c15-18(16)17-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)17/h1-10H

InChI key

FWQDVFJXVXLJCK-UHFFFAOYSA-N

Application

Phosphorous precursor for ligand synthesis[1][2].

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Drinkel, E., et al.
Organometallics, 29, 2503-2514 (2010)
Mariz, R., et al.
Organometallics, 27, 6605-6613 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service