744468
Dibenzylideneacetone palladium(0) phosphaadamantane ethyl Silica
≥45 μm particle size (95%)
Synonym(s):
PhosphonicS™ L-PAPd2r
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
form
powder
extent of labeling
0.01-0.030 mmol/g loading
particle size
≥45 μm (95%)
General description
Dibenzylideneacetone palladium(0) phosphaadamantane ethyl Silica is a silica supported heterogeneous palladium catalyst containing a phosphatrioxoadamantane ligand that can be used for Suzuki cross-coupling.
Application
Silica Supported Cross-Coupling Catalyst
Analysis Note
palladium: detected
Legal Information
Sold in collaboration with PhosphonicS Ltd. for research purposes only.
PhosphonicS is a trademark of PhosphonicS Ltd.
signalword
Warning
hcodes
Hazard Classifications
Skin Sens. 1
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Silicaethylphosphatrioxaadamantane?A new support for palladium catalysts and evaluation in Suzuki coupling reactions
Guino M, et al.
J. Mol. Catal. A: Chem., 293(1), 25-30 (2008)
Guino, M.; et al.
J. Mol. Catal. A: Chem., 293, 25-25 (2008)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service