Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

744298

Sigma-Aldrich

(5R)-5-(1H-Indol-3-ylmethyl)-2,2,3-trimethyl-4-imidazolidinone monohydrochloride

≥98.5% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19N3O · HCl
CAS Number:
Molecular Weight:
293.79
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

≥98.5% (HPLC)
98.5-101.5% (AT)

form

solid

optical activity

[α]/D +106 to +105°, c = 1.43 in methanol

suitability

conforms to structure for Infrared spectrum

SMILES string

Cl.CN1C(=O)[C@@H](Cc2c[nH]c3ccccc23)NC1(C)C

InChI

1S/C15H19N3O.ClH/c1-15(2)17-13(14(19)18(15)3)8-10-9-16-12-7-5-4-6-11(10)12;/h4-7,9,13,16-17H,8H2,1-3H3;1H/t13-;/m1./s1

InChI key

YYTZVVHQTSYLLZ-BTQNPOSSSA-N

Application

Organocatalyst for highly enantioselective Diels-Alder reactions.

Legal Information

U.S. Pat. 6,369,243 and related patents apply. For research purposes only.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guilong Li et al.
Angewandte Chemie (International ed. in English), 52(17), 4628-4632 (2013-03-23)
Mild Magic: A mild, enantioselective Diels-Alder reaction, catalyzed by a chiral magnesium phosphate species, has been developed for the synthesis of various chiral spirooxindoles. Molecular sieves were found to have a considerable effect when used as additives in this reaction.
Asymmetric Intramolecular Aza?Michael Reaction Using Environmentally Friendly Organocatalysis.
Takasu, K., et al.
Heterocycles, 59, 51-55 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service