Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

744220

Sigma-Aldrich

Dichlorobis(dicyclohexyl-1-piperidinylphosphine)palladium(II)

≥97.0% (CHN)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H64Cl2N2P2Pd
CAS Number:
Molecular Weight:
740.16
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

≥97.0% (CHN)

form

powder

composition

carbon, 53.5-56.8%

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

suitability

conforms to structure for Infrared spectrum

SMILES string

Cl[Pd]Cl.C1CCC(CC1)P(C2CCCCC2)N3CCCCC3.C4CCC(CC4)P(C5CCCCC5)N6CCCCC6

InChI

1S/2C17H32NP.2ClH.Pd/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;;/h2*16-17H,1-15H2;2*1H;/q;;;;+2/p-2

InChI key

DGAATSPGVXMFKV-UHFFFAOYSA-L

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeanne L Bolliger et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(36), 11072-11081 (2010-08-18)
[Pd(Cl)(2){P(NC(5)H(10))(C(6)H(11))(2)}(2)] (1) has been prepared in quantitative yield by reacting commercially available [Pd(cod)(Cl)(2)] (cod=cyclooctadiene) with readily prepared 1-(dicyclohexylphosphanyl)piperidine in toluene under N(2) within a few minutes at room temperature. Complex 1 has proved to be an excellent Negishi catalyst, capable
Roman Gerber et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(29), 8901-8905 (2012-06-16)
Cis all round: Dichlorobis[1-(dicyclohexylphosphanyl)piperidine]palladium, [(P{(NC(5)H(10))(C(6)H(11))(2)})(2)Pd(Cl)(2)], is a highly efficient alkyne hydrothiolation catalyst and the first generally applicable system that selectively generates cis-configured anti-Markovnikov adducts in excellent yields within only a few minutes at 120 °C in the presence of only 0.05 mol %
Access to 2-Aminopyridines - Compounds of Great Biological and Chemical Significance
Bolliger, J. L.; et al.
Advanced Synthesis & Catalysis, 353, 945-945 (2011)
Jeanne L Bolliger et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(13), 4075-4081 (2010-02-27)
Dichloro-bis(aminophosphine) complexes are stable depot forms of palladium nanoparticles and have proved to be excellent Suzuki-Miyaura catalysts. Simple modifications of the ligand (and/or the addition of water to the reaction mixture) have allowed their formation to be controlled. Dichlorobis[1-(dicyclohexylphosphanyl)piperidine]palladium (3)
Mizoroki-Heck reactions catalyzed by dichloro{bis[1-(dicyclohexylphosphanyl)piperidine]}palladium. Palladium nanoparticle formation promoted by (water induced) ligand degradation.
Oberholzer, M., et al.
Advanced Synthesis & Catalysis, 354, 627-627 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service