Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

743879

Sigma-Aldrich

HThr(OtBu)-HMPB-ChemMatrix® resin

extent of labeling: 0.30-0.60 mmol/g loading

Synonym(s):

4-{4-[(O-tert-Butyl-L-α-threonyloxy)methyl]-3-methoxyphenoxy}butyric acid, polymer-bound to ChemMatrix® resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

form

powder

extent of labeling

0.30-0.60 mmol/g loading

particle size

35-100 mesh (wet sieved)

application(s)

peptide synthesis

storage temp.

2-8°C

Application

HThr(OtBu)-HMPB-ChemMatrix® resin may be used in solid-phase peptide synthesis.[1][2]

Legal Information

ChemMatrix is a registered trademark of Matrix Innovation, Inc.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ChemMatrix? for complex peptides and combinatorial chemistry.
Garcia?Ramos Y, et al.
The Journal of Peptide Research, 16(12), 675-678 (2010)
ChemMatrix, a poly (ethylene glycol)-based support for the solid-phase synthesis of complex peptides
Garcia-Martin F, et al.
Journal of Combinatorial Chemistry, 8(2), 213-220 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service