Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

743798

Sigma-Aldrich

Chromium(III) ethyl/butyl phosphonate Silica

≥45 μm particle size (95%)

Synonym(s):

PhosphonicS POCr

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.22

form

powder

extent of labeling

0.2-0.5 mmol/g loading

particle size

≥45 μm (95%)

Application

Silica Supported Oxidation Catalyst

Analysis Note

chromium: detected

Legal Information

Sold in collaboration with PhosphonicS Ltd. for research purposes only.
PhosphonicS is a trademark of PhosphonicS Ltd.

signalword

Warning

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Muta. 2 - Ox. Sol. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service