Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

743739

Sigma-Aldrich

2H,6H-[1,2]Iodoxolo[4,5,1-hi][1,2]benziodoxole-2,6-dione

96.0-104.0 wt. % (T)

Synonym(s):

2aλ3-Ioda-2,3-dioxa-cyclopenta[cd]indene-1,4-dione, Iodosodilactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H3IO4
CAS Number:
Molecular Weight:
290.01
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

96.0-104.0% (wt., T)

form

solid

functional group

iodo

SMILES string

O=C1O[I]2OC(=O)c3cccc1c23

InChI

1S/C8H3IO4/c10-7-4-2-1-3-5-6(4)9(12-7)13-8(5)11/h1-3H

InChI key

DHIWEENQJCGNME-UHFFFAOYSA-N

Application

2H,6H-[1,2]Iodoxolo[4,5,1-hi][1,2]benziodoxole-2,6-dione is an iodine(III) reagent that can be used:
  • As a coupling reagent in many organic reactions like amidation, peptide coupling, and esterification to yield amides, esters, peptides, and macrocyclic lactones without racemization.[1]
  • As an oxidant in the arylation reactions of fluorophenols.[2]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Tian et al.
Organic letters, 14(12), 3020-3023 (2012-06-08)
A hypervalent iodine(III) reagent plays a novel role as an efficient coupling reagent to promote the direct condensation between carboxylic acids and alcohols or amines to provide esters, macrocyclic lactones, amides, as well as peptides without racemization. The regeneration of
Recyclable hypervalent iodine (III) reagent iodosodilactone as an efficient coupling reagent for direct esterification, amidation, and peptide coupling
Tian J, et al.
Organic Letters, 14(12), 3020-3023 (2012)
Regioselective oxidative arylation of fluorophenols
Yu C and Patureau FW
Angewandte Chemie (International Edition in English), 58(51), 18530-18534 (2019)
Synthesis and properties of two iodosolactones.
Agosta, W. C.
Tetrahedron Letters, 6, 2681-2685 (1965)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service