Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

743720

Sigma-Aldrich

5-Ethyl-2-methylpyridine

≥96.0% (GC)

Synonym(s):

5-Ethyl-2-picoline, NSC 1984

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11N
CAS Number:
Molecular Weight:
121.18
Beilstein/REAXYS Number:
109269
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

4.2 (vs air)

description

Lonza quality

assay

≥96.0% (GC)

form

liquid

refractive index

n20/D 1.496-1.498
n20/D 1.497 (lit.)

bp

178 °C (lit.)

density

0.919 g/mL at 25 °C (lit.)

SMILES string

CCc1ccc(C)nc1

InChI

1S/C8H11N/c1-3-8-5-4-7(2)9-6-8/h4-6H,3H2,1-2H3

InChI key

NTSLROIKFLNUIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service