Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

743674

Sigma-Aldrich

1,3-Bis(2,7-diisopropylnaphthalen-1-yl)imidazolinium tetrafluoroborate

≥95% (HPLC)

Synonym(s):

2,7-SIPrNap·HBF4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H43BF4N2
CAS Number:
Molecular Weight:
578.53
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

≥95% (HPLC)

form

powder

reaction suitability

reagent type: catalyst

SMILES string

F[B-](F)(F)F.CC(C)c1ccc2ccc(C(C)C)c(N3CC[N+](=C3)c4c(ccc5ccc(cc45)C(C)C)C(C)C)c2c1

InChI

1S/C35H43N2.BF4/c1-22(2)28-11-9-26-13-15-30(24(5)6)34(32(26)19-28)36-17-18-37(21-36)35-31(25(7)8)16-14-27-10-12-29(23(3)4)20-33(27)35;2-1(3,4)5/h9-16,19-25H,17-18H2,1-8H3;/q+1;-1

InChI key

JHTIXBSEIAAUNV-UHFFFAOYSA-N

General description

Mixture of anti and syn isomers in variable ratios. Generally anti as major component.

application

Carbene ligand precursor

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vieille-Petit, L.; et al.
Chemical Communications (Cambridge, England), 3783-3783 (2009)
Xinjun Luan et al.
Journal of the American Chemical Society, 130(21), 6848-6858 (2008-05-01)
A new class of easily accessible and stable imidazolin-2-ylidenes has been synthesized where the side chains are comprised of substituted naphthyl units. Introduction of the naphthyl groups generates C 2 -symmetric ( rac) and C s- symmetric ( meso) atropisomers
Michele Gatti et al.
Journal of the American Chemical Society, 131(27), 9498-9499 (2009-06-19)
Two saturated N-heterocyclic carbene ligands with substituted naphthyl side chains were used for the preparation of Blechert-type ruthenium metathesis precatalysts. The resulting conformers of the complexes were separated and unambiguously assigned by X-ray diffraction studies. All new complexes were compared
Vieille-Petit, L.; et al.
European Journal of Inorganic Chemistry, 13, 1861-1861 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service