Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

743631

Sigma-Aldrich

(4-Iodophenyl)(2,4,6-trimethylphenyl)iodonium triflate

≥98% (HPLC)

Synonym(s):

(4-Iodophenyl)(2,4,6-trimethylphenyl)iodonium trifluoromethanesulfonate, (4-Iodphenyl)mesityliodonium triflate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15F3I2O3S
CAS Number:
Molecular Weight:
598.16
Beilstein/REAXYS Number:
20041342
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

assay

≥98% (HPLC)

form

powder

composition

carbon, 31.3-32.9% CHN

SMILES string

[O-]S(=O)(=O)C(F)(F)F.Cc1cc(C)c([I+]c2ccc(I)cc2)c(C)c1

InChI

1S/C15H15I2.CHF3O3S/c1-10-8-11(2)15(12(3)9-10)17-14-6-4-13(16)5-7-14;2-1(3,4)8(5,6)7/h4-9H,1-3H3;(H,5,6,7)/q+1;/p-1

InChI key

GWJFLDHKPRIPGZ-UHFFFAOYSA-M

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service