Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

742651

Sigma-Aldrich

P,P-Dichloroferrocenylphosphine

≥95% (T)

Synonym(s):

(Dichlorophosphino)ferrocene, FcPCl2, Ferrocenyl dichlorophosphane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9Cl2FeP
CAS Number:
Molecular Weight:
286.90
Beilstein/REAXYS Number:
16508371
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

assay

≥95% (T)

form

solid

reaction suitability

core: iron
reagent type: catalyst
reagent type: ligand

functional group

phosphine

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.ClP(Cl)[C]2[CH][CH][CH][CH]2

InChI

1S/C5H4Cl2P.C5H5.Fe/c6-8(7)5-3-1-2-4-5;1-2-4-5-3-1;/h1-4H;1-5H;

InChI key

XKDGYTLBVLFALN-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

supp_hazards

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service