Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

742368

Sigma-Aldrich

Di-μ-chlorobis[1-[(1R)-1-(dimethylamino)ethyl]-2-naphthyl-C,N]dipalladium(II)

≥97% (HPLC)

Synonym(s):

(R)-N,N-α-Trimethyl-1-naphthalenemethanamine palladium complex, Di-μ-chlorobis[1-[(1R)-1-(dimethylamino-κN)ethyl]-2-naphthalenyl-κC]dipalladium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H32Cl2N2Pd2
CAS Number:
Molecular Weight:
680.31
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

assay

≥97% (HPLC)

form

solid

optical activity

[α]/D -170±5°, c = 0.49 in dichloromethane

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

impurities

1 mol/mol dichlormethane

SMILES string

C[C@@H]1c2c(ccc3ccccc23)[Pd]4([Cl][Pd]5([Cl]4)c6ccc7ccccc7c6[C@@H](C)[N]5(C)C)[N]1(C)C

InChI

1S/2C14H16N.2ClH.2Pd/c2*1-11(15(2)3)13-10-6-8-12-7-4-5-9-14(12)13;;;;/h2*4-9,11H,1-3H3;2*1H;;/t2*11-;;;;/m11..../s1

InChI key

DSNXLWIMELRETE-KGFKVAEWSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service