Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

742139

Sigma-Aldrich

(1R,2R)-trans-2-Aminocycloheptanol

≥99% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NO
CAS Number:
Molecular Weight:
129.20
Beilstein/REAXYS Number:
2070909
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

assay

≥99% (GC)

form

crystals

optical purity

enantiomeric excess: ≥99.5% (GC)

functional group

hydroxyl

SMILES string

N[C@@H]1CCCCC[C@H]1O

InChI

1S/C7H15NO/c8-6-4-2-1-3-5-7(6)9/h6-7,9H,1-5,8H2/t6-,7-/m1/s1

InChI key

ZWOFTVSNNFYPAB-RNFRBKRXSA-N

Application

(1R,2R)-trans-2-Aminocycloheptanol can be used as a reactant for the preparation of 4,6-difluoro-N-(1S,2S)-2-hydroxycyclohexyl-1-((6-(1-methyl-1H-pyrazol-4-yl)pyridine-3-yl)methyl)-1H-indole-3-carboxamide as a potent M1 ago-positive allosteric modulator (PAM).[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diverse effects on M1 signaling and adverse effect liability within a series of M1 Ago-PAMs
Rook JM, et al.
ACS Chemical Neuroscience, 8(4), 866-883 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service