Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

741787

Sigma-Aldrich

3-Thioanisolemagnesium bromide solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7BrMgS
Molecular Weight:
227.40
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

solution

reaction suitability

reaction type: Grignard Reaction

concentration

0.5 M in THF

density

0.951 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CSc1cccc([Mg]Br)c1

InChI

1S/C7H7S.BrH.Mg/c1-8-7-5-3-2-4-6-7;;/h2-3,5-6H,1H3;1H;/q;;+1/p-1

InChI key

KWVGKDRKCDWKAE-UHFFFAOYSA-M

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-0.4 °F

flash_point_c

-18 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service