Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

741760

Sigma-Aldrich

4-Ethoxyphenylmagnesium bromide solution

1 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9BrMgO
CAS Number:
Molecular Weight:
225.37
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solution

reaction suitability

reaction type: Grignard Reaction

concentration

1 M in THF

density

1.004 g/mL at 25 °C

SMILES string

CCOc1ccc([Mg]Br)cc1

InChI

1S/C8H9O.BrH.Mg/c1-2-9-8-6-4-3-5-7-8;;/h4-7H,2H2,1H3;1H;/q;;+1/p-1

Inchi Key

LSLTUAIHOGSOKB-UHFFFAOYSA-M

signalword

Danger

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

1.4 °F

flash_point_c

-17 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service