Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

740993

Sigma-Aldrich

Antimony(III) telluride

beads, ≥99.99%

Synonym(s):

Antimony sesquitelluride, Antimony telluride, Diantimony tritelluride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sb2Te3
CAS Number:
Molecular Weight:
626.32
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

≥99.99%

form

beads

density

6.5 g/mL at 25 °C (lit.)

SMILES string

[Te]=[Sb][Te][Sb]=[Te]

InChI

1S/2Sb.3Te

Inchi Key

BPDQXJZWVBPDSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Antimony(III) telluride (Sb2Te3) is a three dimensional topological insulator that can be used as a binary sesquichalogenide. It forms p-type semiconducting films for the formation of thermoelectric materials.[1][2]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Sb2Te3 in combination with bismuth(III) telluride (Bi2Te3) can form super-lattices, which facilitate the fabrication of devices such as thermoelectric generators.[3][4]

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenzhong Wang et al.
Journal of the American Chemical Society, 127(40), 13792-13793 (2005-10-06)
We report a high-yield synthesis of single-crystal hexagonal Sb2Te3 nanoplates using a solvothermal approach. The experimental results show that the concentration of capping agent CTAB played a key role in the formation of nanoplates. This new nanostructure may provide opportunity
T.C. Harman, B. Paris, S.E. Miller, H.L. Goering
Journal of Physics and Chemistry of Solids, 2, 181-181 (1957)
Chen, J.; Sun, T.; Sim, D. H.; Peng, H.; Wang, H.; Fan, S.;
Chemistry of Materials, 22, 3086-3086 (2010)
A wearable thermoelectric generator fabricated on a glass fabric
Kim SJ, et al.
Energy & Environmental Science, 7(6), 1959-1965 (2014)
Observation of time-reversal-protected single-Dirac-cone topological-insulator states in Bi 2 Te 3 and Sb 2 Te 3
Hsieh D, et al.
Physical Review Letters, 103(14), 146401-146401 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service