Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

740934

Sigma-Aldrich

Xylitol-13C5

99 atom % 13C, 98% (CP)

Synonym(s):

Xylite-13C5

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO13CH2[13CH(OH)]313CH2OH
Molecular Weight:
157.11
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

isotopic purity

99 atom % 13C

assay

98% (CP)

mp

94-97 °C (lit.)

mass shift

M+5

SMILES string

O[13CH2][13CH](O)[13CH](O)[13CH](O)[13CH2]O

InChI

1S/C5H12O5/c6-1-3(8)5(10)4(9)2-7/h3-10H,1-2H2/i1+1,2+1,3+1,4+1,5+1

InChI key

HEBKCHPVOIAQTA-CVMUNTFWSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service