Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

740918

Sigma-Aldrich

Barium tetrafluoronickelate

99.9% trace metals basis

Synonym(s):

Barium nickel fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BaNiF4
CAS Number:
Molecular Weight:
272.01
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
assay:
99.9% trace metals basis
form:
powder

assay

99.9% trace metals basis

form

powder

impurities

≤2000.0 ppm Trace Metal Analysis

mp

>500 °C

SMILES string

[Ba++].F[Ni--](F)(F)F

InChI

1S/Ba.4FH.Ni/h;4*1H;/q+2;;;;;+2/p-4

Inchi Key

VWKLKTCOYQZGPF-UHFFFAOYSA-J

Pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Carc. 1B - Skin Sens. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J F Scott
Science (New York, N.Y.), 315(5814), 954-959 (2007-02-17)
Long viewed as a topic in classical physics, ferroelectricity can be described by a quantum mechanical ab initio theory. Thin-film nanoscale device structures integrated onto Si chips have made inroads into the semiconductor industry. Recent prototype applications include ultrafast switching
Ederer, C. and Spaldin, N. A.
Physical Review. B, Condensed Matter and Materials Physics, 74, 024102-024102 (2006)
DiDomenico, Jr. M.; Eibshutz, M.; Guggenheim, H. J.; Camlibel, I.
Solid State Communications, 7, 1119-1119 (1969)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service