Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

740276

Sigma-Aldrich

3-Benzyloxypropionaldehyde

96%

Synonym(s):

3-Benzyloxy-1-propanal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

96%

form

liquid

refractive index

n20/D 1.512

density

1.059 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

O=CCCOCc1ccccc1

InChI

1S/C10H12O2/c11-7-4-8-12-9-10-5-2-1-3-6-10/h1-3,5-7H,4,8-9H2

InChI key

ANSYAMHYCYOWAW-UHFFFAOYSA-N

Caution

Handle under nitrogen or inert gas.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service