Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

740101

Sigma-Aldrich

N-Boc-2-bromo-3-methylindole

97%

Synonym(s):

1-(tert-Butyloxycarbonyl)-2-bromo-3-methylindole, 2-Bromo-3-methyl-indole-1-carboxylic acid tert-butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16BrNO2
CAS Number:
Molecular Weight:
310.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

160-170 °C

functional group

bromo

storage temp.

2-8°C

SMILES string

Cc1c(Br)n(C(=O)OC(C)(C)C)c2ccccc12

InChI

1S/C14H16BrNO2/c1-9-10-7-5-6-8-11(10)16(12(9)15)13(17)18-14(2,3)4/h5-8H,1-4H3

InChI key

NVJYNNQGUPOOCB-UHFFFAOYSA-N

pictograms

Skull and crossbonesEnvironment

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service