Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

740098

Sigma-Aldrich

1,1-Dibromo-2-vinylcyclopropane

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6Br2
CAS Number:
Molecular Weight:
225.91
UNSPSC Code:
12352100
PubChem Substance ID:

assay

96%

form

liquid

refractive index

n20/D 1.547

density

1.869 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

BrC1(Br)CC1C=C

InChI

1S/C5H6Br2/c1-2-4-3-5(4,6)7/h2,4H,1,3H2

InChI key

RTBUVGWEJCPSFF-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service