Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

739561

Sigma-Aldrich

Thallium(I) trifluoromethanesulfonate

97%

Synonym(s):

Thallium(I) trifluoromethylsulfonate, Tl(OTf)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CF3O3STl
CAS Number:
Molecular Weight:
353.45
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

powder

reaction suitability

core: thallium
reagent type: catalyst

mp

245-250 °C

storage temp.

2-8°C

SMILES string

[Tl+].[O-]S(=O)(=O)C(F)(F)F

InChI

1S/CHF3O3S.Tl/c2-1(3,4)8(5,6)7;/h(H,5,6,7);/q;+1/p-1

InChI key

PDFSYPYWCONFDC-UHFFFAOYSA-M

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel T Walters et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(15), 3849-3857 (2018-12-24)
Two new molecular boxes, the mono-bromo box [Au6 (Triphos)4 Br](SbF6 )5 ⋅6(CH2 Cl2 ), 4 mB, and the dibromo box, [Au6 (Triphos)4 Br2 ⋅H2 O](SbF6 )4 ⋅4(CH2 Cl2 ), 5 dB, have been prepared in crystalline form. Although constructed from non-luminescent components

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service