Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

739359

Sigma-Aldrich

S-(10-Undecenyl) thioacetate

97%

Synonym(s):

10-Undecenyl 1-thioacetate, Thioacetic acid, S-(10-undecenyl) ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CH(CH2)8CH2SC(O)CH3
CAS Number:
Molecular Weight:
228.39
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

assay

97%

form

liquid

refractive index

n20/D 1.475

density

0.906 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(=O)SCCCCCCCCCC=C

InChI

1S/C13H24OS/c1-3-4-5-6-7-8-9-10-11-12-15-13(2)14/h3H,1,4-12H2,2H3

InChI key

SHNFVLZZKWMGSP-UHFFFAOYSA-N

Application

This self-assembly material is used as a precursor in the creation of a multifunctional molecular layer as a junction between metallic zinc and polymer coatings such as polydimethylsiloxane (PDMS).[1]

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Berger, F.; Delhalle, J.; Mekhalif, Z.
Electrochimica Acta, 54, 6464-6464 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service