Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

739014

Sigma-Aldrich

Bis[(2-diisopropylphosphino]ethyl)amine solution

10 wt. % in THF

Synonym(s):

Bis((2-diisopropylphosphino)ethyl)-amine, Bis[(2-di-i-propylphosphino]ethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H37NP2
Molecular Weight:
305.42
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

10 wt. % in THF

refractive index

n/D 1.418

density

0.884 g/mL at 25 °C

SMILES string

CC(C)P(CCNCCP(C(C)C)C(C)C)C(C)C

InChI

1S/C16H37NP2/c1-13(2)18(14(3)4)11-9-17-10-12-19(15(5)6)16(7)8/h13-17H,9-12H2,1-8H3

InChI key

FTVIGQGOGIHMBS-UHFFFAOYSA-N

Legal Information

Sold under license from Kanata for research purposes only.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system, Respiratory system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

1.4 °F

flash_point_c

-17.0 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service