Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

738735

Sigma-Aldrich

Poly(N-isopropylacrylamide-co-acrylamide)

acrylamide 22-25 mol %

Synonym(s):

Acrylamide copolymer, Poly(NIPAM-co-acrylamide)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11NO)x (C3H5NO)y
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

mol wt

average Mn 20,000-25,000

composition

acrylamide, 22-25 mol %

transition temp

Tg 154-159 °C
critical temperature 30-34 °C

Mw/Mn

1.6

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

By altering the physicochemical properties, smart or intelligent drug delivery systems can be designed to deliver therapeutic molecules on-demand. Learn more about the application of stimuli-responsive materials in drug delivery.

Immunosuppressive tumor-associated myeloid cells (TAMC) are responsible for glioblastoma (GBM) resistance to immunotherapies and existing standard of care treatments. This mini-review highlights recent progress in implementing nanotechnology in advancing TAMC-targeted therapies for GBM.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service