Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

737801

Sigma-Aldrich

Fmoc-Gly-Bt

97%

Synonym(s):

9H-Fluoren-9-yl)methyl 2-(1H-benzo[d][1,2,3]triazol-1-yl)-2-oxoethylcarbamate, N-Fmoc-(1-benzotriazolylcarbonyl)methylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H18N4O3
CAS Number:
Molecular Weight:
398.41
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

assay

97%

form

solid

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

160-168 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

−20°C

SMILES string

O=C(NCC(=O)n1nnc2ccccc12)OCC3c4ccccc4-c5ccccc35

InChI

1S/C23H18N4O3/c28-22(27-21-12-6-5-11-20(21)25-26-27)13-24-23(29)30-14-19-17-9-3-1-7-15(17)16-8-2-4-10-18(16)19/h1-12,19H,13-14H2,(H,24,29)

InChI key

FKDIZKBMJXDKNM-UHFFFAOYSA-N

Application

Benzotriazole amino acids, or aminoacylbenzotriazolides, are versatile reagents for synthesizing peptides as well as their mimetics and conjugates. Benzotriazole amino acids have been used for the preparation of diverse derivatives including:

  • Polypeptidal benzotriazolides[1]
  • Peptidomimetics, such as aminoxypeptides[2], depsipeptides[3] and heterocyclic peptidomimetics[4]
  • Tagged peptides and peptidomimetics, particularly those with fluorescent labels[5][6]
  • N, O, S, and C linked peptide conjugates[6]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katritzky, A. R.; Narindoshvili, T.; Angrish, P.
Synthesis, 2013-2022 (2008)
Hansen, F. K.; Beagle, L. K.; Todadze, E.; Katritzky, A. R.
Heterocycles, 515-526 (2012)
Abdelmajeid, A.; Tala, S. R.; Amine, M. S.; Katritzky, A. R.
Synthesis, 2995-3005 (2011)
Katritzky, A. R.; Angrish, P.; Todadze, E.
Synlett, 2392-2411 (2009)
Ilker Avan et al.
The Journal of organic chemistry, 76(12), 4884-4893 (2011-04-02)
Reactions of O-Pg(α-hydroxyacyl)benzotriazoles with (a) unprotected α-hydroxycarboxylic acids, (b) amino acids, and (c) amines afforded, respectively, chirally pure (a) oligoesters, (b) depsidipeptides, and (c) amide conjugates (yields 52-94%). N-Pg(α-Aminoacyl)benzotriazoles reacted with α-hydroxycarboxylic acids to yield depsidipeptides (47-87%). N-Pg(depsidipeptidoyl)benzotriazoles, obtained from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service