Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

737666

Sigma-Aldrich

(S,S)-cis EZ-CrotylMix

Synonym(s):

(25:1)-(3aS,7aS)-1,3-Bis(4-bromobenzyl)-2-((Z)-but-2-enyl)-2-chlorooctahydro-1H-benzo[d][1,3,2]diazasilole:scandium(III) triflate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29Br2ClF9N2O9S3ScSi
Molecular Weight:
1061.01
UNSPSC Code:
12352103
PubChem Substance ID:

form

powder

Quality Level

SMILES string

FC(F)(F)S(=O)(=O)O[Sc](OS(=O)(=O)C(F)(F)F)OS(=O)(=O)C(F)(F)F.C\C=C/C[Si]1(Cl)N(Cc2ccc(Br)cc2)[C@H]3CCCC[C@@H]3N1Cc4ccc(Br)cc4

InChI

1S/C24H29Br2ClN2Si.3CHF3O3S.Sc/c1-2-3-16-30(27)28(17-19-8-12-21(25)13-9-19)23-6-4-5-7-24(23)29(30)18-20-10-14-22(26)15-11-20;3*2-1(3,4)8(5,6)7;/h2-3,8-15,23-24H,4-7,16-18H2,1H3;3*(H,5,6,7);/q;;;;+3/p-3/b3-2-;;;;/t23-,24-;;;;/m0..../s1

InChI key

PRMHXJZNOJQQCI-QGJQGQTRSA-K

Legal Information

EZ-CrotylMix is a trademark of Columbia University

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service