Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

737623

Sigma-Aldrich

Z-val-bt

95%

Synonym(s):

(S)-N-Z-1-Benzotriazolylcarbonyl-2-methylpropylamine, (S)-Benzyl 1-(1H-benzo[d][1,2,3]triazol-1-yl)-3-methyl-1-oxobutan-2-ylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20N4O3
CAS Number:
Molecular Weight:
352.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

110-115 °C

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

O=C([C@@H](NC(OCC1=CC=CC=C1)=O)C(C)C)N(N=N2)C3=C2C=CC=C3

InChI

1S/C19H20N4O3/c1-13(2)17(20-19(25)26-12-14-8-4-3-5-9-14)18(24)23-16-11-7-6-10-15(16)21-22-23/h3-11,13,17H,12H2,1-2H3,(H,20,25)/t17-/m0/s1

InChI key

HXOSDDZLLFFPFC-KRWDZBQOSA-N

Application

Benzotriazole amino acids, or aminoacylbenzotriazolides, are versatile reagents for synthesizing peptides as well as their mimetics and conjugates. Benzotriazole amino acids have been used for the preparation of diverse derivatives including:

  • Polypeptidal benzotriazolides[1]
  • Peptidomimetics, such as aminoxypeptides[2], depsipeptides[3] and heterocyclic peptidomimetics[4]
  • Tagged peptides and peptidomimetics, particularly those with fluorescent labels[5],[6]
  • N, O, S, and C linked peptide conjugates[6]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katritzky, A. R.; et al.
The Journal of Organic Chemistry, 8690-8694 (2009)
Hansen, F. K.; et al.
Heterocycles, 515-526 (2012)
Katritzky, A. R.; et al.
Synthesis, 2013-2022 (2008)
Katritzky, A. R.; et al.
Synlett, 2392-2411 (2009)
Abdelmajeid, A.; et al.
Synthesis, 2995-3005 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service