Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

737437

Sigma-Aldrich

Magnesium chloride triethylamine solution

0.5 M in methanol

Synonym(s):

MgCl2-Et3N Base System

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15N · MgCl2
Molecular Weight:
196.40
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.22

form

solution

concentration

0.5 M in methanol

density

0.850 g/mL at 25 °C

SMILES string

Cl[Mg]Cl.CCN(CC)CC

InChI

1S/C6H15N.2ClH.Mg/c1-4-7(5-2)6-3;;;/h4-6H2,1-3H3;2*1H;/q;;;+2/p-2

InChI key

FMUNKBAIXLTEGC-UHFFFAOYSA-L

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 1 - STOT SE 3

target_organs

Eyes,Central nervous system, Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

48.2 °F - closed cup

flash_point_c

9 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service